Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

00596.FF
00596.FFEA 185 EUR
00596.FF
Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Holmium

Purity: 99.9% CAS No.: 7440-60-0 Physical Form: ≈3.4g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011049

Formule: Ho
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011049
CAS nummer: 7440-60-0
EINECS: 231-169-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR