Lutetium ≥99.9% (REO, rare earth oxide basis), pieces

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

00628.03 00628.06
00628.03EA 245 EUR
00628.03 00628.06
Lutetium ≥99.9% (REO, rare earth oxide basis), pieces
Lutetium

Purity: 99.9% CAS No.: 7439-94-3 Physical Form: Packaged in air-stable glass bottle Odour: Odourless MDL No.: MFCD00011098

Formule: Lu
Kookpunt: 3000 °C (1013 hPa)
Smeltpunt: 1651…1675 °C
Dichtheid: 9,849 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011098
CAS nummer: 7439-94-3
EINECS: 231-103-0
Merck Index: 12,05642

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR