Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

00653.06 00653.18 00653.30
00653.06EA 77.2 EUR
00653.06 00653.18 00653.30
Barium ≥99.2% (metals basis), pieces ≤ 2.5 cm (1 in) (max. 0.8% Sr)
Barium

Purity: 99.2% CAS No.: 7440-39-3 Physical Form: Packaged in oil Appearance: Yellow-silver Odour: Odourless UN No.: 1400 MDL No.: MFCD00134031

Formule: Ba
Kookpunt: 1640 °C (1013 hPa)
Smeltpunt: 725 °C
Dichtheid: 3,51 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134031
CAS nummer: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR