Silver ≥99.9% (metals basis), spherical powder -325 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

00678.09 00678.18
00678.09EA 110 EUR
00678.09 00678.18
Silver ≥99.9% (metals basis), spherical powder -325 mesh
Silver

Purity: 99.9% CAS No.: 7440-22-4 Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397

Formule: Ag
Kookpunt: 2212 °C (1013 hPa)
Dichtheid: 10,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR