Tin ≥99.8% (metals basis), ingot 2.54x0.95x30.5 cm (1.0x0.37x12.0 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

00929.KF 00929.KJ
00929.KFEA 117 EUR
00929.KF 00929.KJ
Tin ≥99.8% (metals basis), ingot 2.54x0.95x30.5 cm (1.0x0.37x12.0 in)
Tin

Purity: 99.8% CAS No.: 7440-31-5 Physical Form: Cast ingot, dimensions are approximate Odour: Odourless MDL No.: MFCD00133862

Formule: Sn
Kookpunt: 2270 °C (1013 hPa)
Smeltpunt: 232 °C
Dichtheid: 7,31 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133862
CAS nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR