Citric acid monohydrate 99.5-100.5% (by anhydrous basis) USP, FCC, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

0119-01EA 97.5 EUR
0119-01 0119-05 0119-07 0119-09
Citric acid monohydrate 99.5-100.5% (by anhydrous basis) USP, FCC, J.T.Baker®
Citric acid monohydrate
Formule: C₆H₈O₇.H₂O
Kookpunt: 280…285 °C (1013 hPa)
Smeltpunt: 37 °C
Dichtheid: 1,54 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149972
CAS nummer: 5949-29-1
EINECS: 231-729-4
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Food GMP Manufactured Product 99.5 - 100.5%
Meets U.S.P Requirements Passes Test
GMP Manufactured Product Passes Test
Meets F.C.C. Requirements Passes Test
Preserve in Tight Containers 7.5 - 9.0%
CAUTION: For Manufacturing, processing or repackaging ≤0.05%
Bulk Food Chemical ≤0.036%
Bulk Pharmaceutical Chemical ≤0.015%
USP - Assay (as C₆H₈O₇) (calculated on anhydrous basis) 99.5 - 100.5 %
USP - Clarity of Solution Passes Test
USP - Color of Solution Passes Test
USP - Identification (USP) Passes Test
USP - Water (H₂O) (U.S.P.) 7.5 - 9.0 %
USP - Residue on Ignition ≤0.05 %
USP - Oxalic Acid (USP) ≤0.036 %
USP - Sulfate (SO₄) ≤0.015 %
USP - Readily Carbonizable Substances Passes Test
FCC - Assay (as C₆H₈O₇) (calculated on anhydrous basis) 99.5 - 100.5 %
FCC - Identification (FCC) Passes Test
FCC - Water (H₂O) (F.C.C.) ≤8.8 %
FCC - Residue on Ignition ≤0.05 %
FCC - Oxalate (C₂O₄) (FCC) ≤100 mg/kg
FCC - Readily Carbonizable Substances Passes Test
FCC - Lead (Pb) ≤0.5 mg/kg
Appearance (white, crystalline powder) Passes Test
Must be subjected to further processing during the preparation of
injectable dosage forms.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR