Potassium chloride, BAKER, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassiumchloride

0209.1000EA 38.04 EUR
0209.1000 0209.5000
Potassium chloride, BAKER, J.T.Baker®
Potassium chloride
Formule: KCl
Kookpunt: 1420 °C (1013 hPa)
Smeltpunt: 773 °C
Dichtheid: 1,98 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Meets Reagent Specifications for testing Ph. Eur. monographs.
Assay (dried substance) 99.0 - 100.5 %
Acidity or Alkalinity passes test
Appearance of solution passes test
Bromides max. 0.1 %
Barium (Ba) passes test
Iron (Fe) max. 20 ppm
Heavy Metals (as Pb) max. 10 ppm
Identification passes test
Iodides passes test
Loss on Drying at 105°C max. 1.0 %
Magnesium and Alkaline-earth Metals (as Ca) max. 200 ppm
Sulfates (as SO4) max. 300 ppm
Additional Specification(s):
Calcium and Magnesium passes test
Sodium (Na) max. 0.1 %
Organic Volatile Impurities passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR