Dimethylsulfoxide ≥99.9% ACS

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

0231-500MLEA 140 EUR
0231-500ML 0231-4L
Dimethylsulfoxide ≥99.9% ACS
Dimethylsulfoxide
Formule: (CH₃)₂SO
MW: 78.14 g/mol
Kookpunt: 189 °C (1013 hPa)
Smeltpunt: 18.5 °C
Dichtheid: 1.101 g/cm³ (20 °C)
Vlampunt: 87
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002089
CAS nummer: 67-68-5
EINECS: 200-664-3
Bereik: 01-2119431362-50
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@270 nm <0.4
Abs.@290 nm ≤0.18
Abs.@310 nm ≤0.06
Abs.@330nm ≤0.02
Abs.@350 – 400 nm ≤0.01
Purity ≥99.9%
Residue After Evaporation ≤0.01%
Titrable Free Acid ≤0.001 meq/g
Water (KF) ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR