ortho-Fosforzuur ≥ 85% ≥85.0-88.0% NF, Multi-Compendial, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Phosphoric acid

Management of Change (MOC) category = R

0268-05EA 396.18 EUR
0268-05 0268-15 0268-00 0268-03 0268-06 0268-07
ortho-Fosforzuur ≥ 85% ≥85.0-88.0% NF, Multi-Compendial, J.T.Baker®
ortho-Fosforzuur

Certificaten: GMP manufactured product, meets NF requirement and EP and BP chemical specification.

Formule: H₃PO₄
Kookpunt: 320 °C (1013 hPa)
Smeltpunt: 75 °C
Dichtheid: 1,69 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011340
CAS nummer: 7664-38-2
EINECS: 215-540-4
UN: 1805
ADR: 8,III
Bereik: 01-2119490790-32
Merck Index: 13,08662

Order Now

Specification Test Results

GMP Manufactured Product Passes Test
Meets E.P. Chemical Specifications Passes Test
Meets B.P. Chemical Specifications Passes Test
Meets N.F. Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤0.001%
NF - Identification Passes Test
NF - Nitrate (NO₃) Passes Test
NF - Phosphorous or Hypophosphorous Acid Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Alkali Phosphates Passes Test
NF - Assay 85.0 - 88.0 %
EP/BP - Assay (H₃PO₄) 84.0 - 90.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Substances Precipitated with Ammonia Passes Test
EP/BP - Phosphorous or Hypophosphorous Acid Passes Test
EP/BP - Chloride (Cl) ≤ 50 ppm
EP/BP - Sulfate (SO₄) ≤ 100 ppm
EP/BP - Iron (Fe) ≤ 50 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR