Natriumchloride, BAKER ANALYZED® ACS, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Saline, NaCl, Sea salt, Halite

0277.1000EA 62.99 EUR
0277.1000 0277.5000 0277.0500 0277.9025PE 0277.9010
Natriumchloride, BAKER ANALYZED® ACS, J.T.Baker®
Natriumchloride
Formule: NaCl
Kookpunt: 1413 °C (1013 hPa)
Smeltpunt: 801 °C
Dichtheid: 2,16 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003477
CAS nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Meets ACS Specifications
Assay min. 99.0 %
Chlorate and Nitrate (as NO3) max. 0.003 %
Barium (Ba) passes test
Bromide (Br) max. 0.01 %
Calcium (Ca) max. 0.002 %
Iodide (I) max. 0.002 %
Potassium (K) max. 0.005 %
Magnesium (Mg) max. 0.001 %
Insoluble Matter max. 0.005 %
pH of 5% Solution at 25°C 5.0 - 9.0
Sulfate (SO4) max. 0.004 %
Trace Impurities (in ppm)
Iron (Fe) max. 2
Heavy Metals (as Pb) max. 5
Phosphate (PO4) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR