Guanidiniumchloride ≥99.5% for biotechnology
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
Synoniemen: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
0287-500GEA 259 EUR
0287-500G
Guanidiniumchloride ≥99.5% for biotechnology
Guanidiniumchloride
Suited for purification of nucleic acids or polypeptides from cell sources and inhibition of both RNase and protease activity.
Formule: NH₂C(=NH)NH₂·HCl
Smeltpunt: 185 °C
Dichtheid: 1,354 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013026
CAS nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Solubility (6 M, Water) Pass
Absorbance 260 nm (6 M, Water) ≤0.03
Arsenic ≤0.0005%
DNase None Detected
Iron ≤0.0005%
Lead <0.0005%
Melting Point 181 - 187 °C
Protease None Detected
Purity >99.5%
RNase None