Vanderzant Modification Vitamin Mixture, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0290324401 0290324405
ICNA0290324401EA 329 EUR
ICNA0290324401 0290324405.
Vanderzant Modification Vitamin Mixture, MP Biomedicals
Dierenbenodigdheden Voeding voor dieren
Vanderzant vitamin mixture for insects is an animal-derived component free vitamin mixture.
  • Mixture of 12 vitamins
  • Powdered
Developed for use in insect cell culture.