Disodium hydrogen phosphate, BAKER ANALYZED® ACS, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0306.0500EA 80.19 EUR
0306.0500 0306.1000 0306.5000
Disodium hydrogen phosphate, BAKER ANALYZED® ACS, J.T.Baker®
Disodium hydrogen phosphate
Formule: Na₂HPO₄
Smeltpunt: 250 °C
Dichtheid: 1,53 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003496
CAS nummer: 7558-79-4
EINECS: 231-448-7
Bereik: 01-2119489797-11
Merck Index: 12,08805

Order Now

Specification Test Results

Meets ACS Specifications
Assay min. 99.0 %
Chloride (Cl) max. 0.002 %
Iron (Fe) max. 0.002 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.01 %
Loss on Drying at 105°C max. 0.2 %
pH of 5% Solution at 25°C 8.7 - 9.3
Sulfate (SO4) max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR