Ammonium sulphate 6% w/v aqueous solution, BAKER ANALYZED®, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

0333-07EA 0 EUR
0333-07
Ammonium sulphate 6% w/v aqueous solution, BAKER ANALYZED®, J.T.Baker®
Ammonium sulphate
Formule: (NH₄)₂SO₄
MW: 132.14 g/mol
Dichtheid: 1.72 g/cm³
MDL Nummer: MFCD00003391
CAS nummer: 7783-20-2
Merck Index: 13,00559

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 5.9 - 6.1%
Appearance Passes Test
Assay ((NH₄)₂SO₄) (by formol method) 5.9 - 6.1 %
pH ("As Is" basis) 5.0 - 6.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR