Kaliumhydroxide ∼45% in aqueous solution for HPLC, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

03564-100ML 03564-500ML
03564-100MLEA 41.7 EUR
03564-100ML 03564-500ML
Kaliumhydroxide ∼45% in aqueous solution for HPLC, Fluka™
Kaliumhydroxide
Formule: KOH
Kookpunt: 112 °C (1,013 hPa)
Smeltpunt: -6 °C
Dichtheid: 1,510 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Assay 40.5 - 49.5 %
Assay of K₂CO₃ Max. 0.3 %
Suitability test (HPLC) complying
Density (D 20/4) 1.440 - 1.480
Absorbance at 500 nm Max. 0.02
Absorbance at 310 nm Max. 0.02
Absorbance at 300 nm Max. 0.03
Absorbance at 290 nm Max. 0.05
Absorbance at 240 nm Max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR