Supelco® PESTANAL® Organic Reference Standard, Diquat Dibromide-(Ethylene-D₄)

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

03627-5MG
03627-5MGEA 193 EUR
03627-5MG
Supelco® PESTANAL® Organic Reference Standard, Diquat Dibromide-(Ethylene-D₄)
Standaarden Organische standaarden

  • GHS signal: GHS06, GHS08, GHS09


Opgelet: P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310 - P305 + P351 + P338 - P314

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d034589ca7a507f42a89c5f561a1d8ab550886f&vid=9d034589ca7a507f42a89c5f561a1d8ab550886f&prid=web&clid=SAPDAM

Order Now

Specificaties

Synonym(s) 1,1′-Ethylene-d₄-2,2′-dipyridylium dibromide
Environmentally Preferable
M-Clarity quality level MQ100
Formule C₁₂D₄H₈Br₂N₂
Moleculair gewicht 348,07
Beschrijving van de gevarenklasse H290 - H301 + H311 - H315 - H317 - H319 - H330 - H335 - H372 - H410
Vlampunt Not applicable
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR