Supelco® PESTANAL® Organic Reference Standard, α-HCH-d₆

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

03811-5MG
03811-5MGEA 859 EUR
03811-5MG
Supelco® PESTANAL® Organic Reference Standard, α-HCH-d₆
Standaarden Organische standaarden

  • GHS signal: GHS06, GHS08, GHS09


Verpakking: Bottomless glass bottle. Contents are inside inserted fused cone.


Opgelet: P202 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P312

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c89df71ec52514936ceb7f546db2c0ee22d0da29&vid=c89df71ec52514936ceb7f546db2c0ee22d0da29&prid=web&clid=SAPDAM

Order Now

Specificaties

M-Clarity quality level MQ100
Synonym(s) α-1,2,3,4,5,6-Hexachlorocyclohexane-d₆; α-BHC-d₆
Environmentally Preferable
CAS No. 86194-41-4
MDL No. MFCD01632293
Formule C₆D₆Cl₆
Moleculair gewicht 296,87
Smeltpuntapparaat 159...160 °C
Beschrijving van de gevarenklasse H301 - H312 - H351 - H410
Vlampunt Not applicable
Shipping conditions STD
Storage conditions 2...8 °C
Opslagtemperatuur 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR