Diammoniumperoxodisulfaat (APS) ≥98% ACS

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

0486-100GEA 79.3 EUR
0486-100G 0486-25G
Diammoniumperoxodisulfaat (APS) ≥98% ACS
Diammoniumperoxodisulfaat (APS)
Formule: (NH₄)₂S₂O₈
Smeltpunt: 120 °C
Dichtheid: 1.98 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003390
CAS nummer: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride & Chlorate <0.001%
Heavy Metals (as Pb) ≤0.005%
Insolubles ≤0.005%
Iron ≤0.001%
Manganese ≤0.00005%
Purity ≥98%
Residue on Ignition ≤0.05%
Titrable Free Acid ≤0.04 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR