Ethidiumbromide 98 Biotechnology

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

0492-25GEA 684 EUR
0492-25G 0492-5G
Ethidiumbromide 98 Biotechnology
Ethidiumbromide

Fluorescent stain for visualizing nucleic acids.


At a concentration of 0.5 μg/ml, ethidium bromide is useful in agarose or acrylamide gels for band detection. It is used to detect as little as 10 ng of nucleic acid. At higher concentrations (0.5 to 1.0 μg/ml), it may be used to facilitate purification of DNA in a cesium chloride gradient.


For maximum convenience, VWR offers ethidium bromide in various forms, including ready-to-use solutions and powdered forms. Each is packaged in shatter-resistant packaging for added safety. VWR’s ethidium bromide dropper bottle offers the ultimate convenience and safety. Simply dispense one drop of ethidium bromide into 50ml of agarose gel or running buffer (TBE or TAE) or staining solution.

Formule: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Kookpunt: 72 °C (0.1 Torr)
Smeltpunt: 261…264 °C
Dichtheid: 1.01 g/cm³ (20 °C)
Vlampunt: <100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011724
CAS nummer: 1239-45-8
EINECS: 214-984-6
UN: 2811
ADR: 6.1 I
Merck Index: 13,04751

Order Now

Specification Test Results

DNase None Detected
Loss on Drying ≤5%
Protease None Detected
Purity (Dry Basis) ≥98%
RNase None


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR