Diammonium hydrogen orthophosphate ≥98% ACS

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

0569-1KGEA 238 EUR
0569-1KG 0569-2.5KG 0569-500G
Diammonium hydrogen orthophosphate ≥98% ACS
Diammonium hydrogen orthophosphate
Formule: (NH₄)₂HPO₄
Smeltpunt: 185 °C
Dichtheid: 1,619 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010891
CAS nummer: 7783-28-0
EINECS: 231-987-8
Bereik: 01-2119490974-22
Merck Index: 13,00546

Order Now

Specification Test Results

Calcium <0.001%
Chloride ≤0.001%
Heavy Metals (as Pb) ≤0.001%
Insolubles ≤0.005%
Iron ≤0.001%
Magnesium ≤0.0005%
Nitrate ≤0.003%
pH (5%, Water) @25 °C 7.7 – 8.1
Potassium ≤0.005%
Purity ≥98%
Sodium ≤0.005%
Sulphate ≤0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR