Natrium-diwaterstoffosfaat, anhydrous ≥98.0%, Reagent Grade

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic

0571-2.5KGEA 506 EUR
0571-2.5KG 0571-500G 0571-100KG
Natrium-diwaterstoffosfaat, anhydrous ≥98.0%, Reagent Grade
Natrium-diwaterstoffosfaat
Formule: NaH₂PO₄
MW: 119.98 g/mol
Smeltpunt: ∼650 °C
Dichtheid: 0.9996 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003527
CAS nummer: 7558-80-7
EINECS: 231-449-2
Bereik: 01-2119489796-13
Merck Index: 12,08806

Order Now

Specification Test Results

Heavy Metals (as Pb) 0.001%
Insolubles = 0.03%
Moisture (KF) = 0.3%
pH (1%, Water) @25°C 4.4 - 4.7
Purity >= 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR