(±)-1,2-Propanediol, high purity

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0575-1LEA 91.2 EUR
0575-1L 0575-4L
(±)-1,2-Propanediol, high purity
(±)-1,2-Propanediol
Formule: CH₃CH(OH)CH₂OH
Kookpunt: 187,6 °C (1013 hPa)
Smeltpunt: –59 °C
Dichtheid: 1,04 g/cm³ (20 °C)
Vlampunt: 99 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064272
CAS nummer: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Arsenic ≤0.0003%
Chloride ≤0.007 %
Heavy metals ≤0.0005%
Purity ≥99.5%
Residue on ignition ≤0.01%
Sulphate ≤0.006%
Water (KF) ≤0.2%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR