Sodium sulphite, anhydrous ≥98% ACS

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0628-1KGEA 175 EUR
0628-1KG 0628-2.5KG 0628-500G
Sodium sulphite, anhydrous ≥98% ACS
Sodium sulphite
Formule: Na₂SO₃
MW: 126.04 g/mol
Smeltpunt: 500 °C
Dichtheid: 2.63 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003503
CAS nummer: 7757-83-7
EINECS: 231-821-4
Bereik: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Chloride <0.02%
Free acid test pass
Heavy metals (as Pb) ≤0.001%
Insolubles ≤0.005%
Iron ≤0.001%
Purity ≥98%
Titrable free base ≤0.03 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR