Magnesium sulphate heptahydrate ACS

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

0662-1KGEA 160 EUR
0662-1KG 0662-2.5KG 0662-500G
Magnesium sulphate heptahydrate ACS
Magnesium sulphate heptahydrate
Formule: MgSO₄·7H₂O
Smeltpunt: 1124 °C
Dichtheid: 2,66 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149785
CAS nummer: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

Ammonium <0.002%
Calcium ≤0.02%
Chloride ≤0.0005%
Heavy metals (as Pb) ≤0.0005%
Insolubles ≤0.005%
Iron ≤0.0005%
Manganese ≤0.0005%
Nitrate ≤0.002%
pH (5%, water) @25 °C 5 – 8.2
Potassium ≤0.005%
Purity 98 – 102%
Sodium ≤0.005%
Strontium ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR