Zwavelzuur 95-97%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

07208-50KG 07208-1L 07208-2.5L 07208-2.5L-GL 07208-4X2.5L 07208-4X5L 07208-5L 07208-6X1L 07208-4X2.5L-G
07208-50KGEA 284 EUR
07208-50KG 07208-1L 07208-2.5L 07208-2.5L-GL 07208-4X2.5L 07208-4X5L 07208-5L 07208-6X1L 07208-4X2.5L-GL
Zwavelzuur 95-97%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Zwavelzuur
Formule: H₂SO₄
Kookpunt: 310 °C (1,013 hPa)
Smeltpunt: –14…–10 °C
Dichtheid: 1,840 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Assay 95.0 - 97.0 %
Residue on ignition Max. 0.0005 %
Arsenic (As) Max. 0.1 ppm
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 2 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Sodium (Na) Max. 0.005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 1 ppm
Nitrate (NO₃) complying
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR