2-Propanol 99 Biotechnology

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

0918-1LEA 73.6 EUR
0918-1L 0918-4L 0918-20L 0918-500ML
2-Propanol 99 Biotechnology
2-Propanol
Formule: (CH₃)₂CHOH
MW: 60.1 g/mol
Kookpunt: 82 °C (1013 hPa)
Smeltpunt: –89 °C
Dichtheid: 0.786 g/cm³ (20 °C)
Vlampunt: 12
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011674
CAS nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
Bereik: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c4eafe19208840993a64d7c98fc917a6f7ccdb1&vid=2cbd35cca07cb89a846356568c2b7828611f7895&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Colour (APHA) ≤10
Density 0.782 - 0.788 g/ml
Moisture (KF) ≤0.2%
Purity >= 99,0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR