Aceton 99,8, SupraSolv® GC-ECD/FID, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-Propanone, DMK, Propanone, Dimethyl ketone

MQ100

1.00012.1000 1.00012.2500 1.00012.4000 1.00012.9030
1.00012.1000EA 70.9 EUR
1.00012.1000 1.00012.2500 1.00012.4000 1.00012.9030
Aceton 99,8, SupraSolv® GC-ECD/FID, Supelco®
Aceton

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionisation Detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formule: CH₃COCH₃
MW: 58.08 g/mol
Kookpunt: 56.2
Smeltpunt: –95.4 °C
Dichtheid: 0.792 g/cm³ (20 °C)
Vlampunt: <–20 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
Bereik: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) =99.8%
Identity(IR) Conforms
Evaporation Residue <= 3.0 mg/L
Water = 0.05%
Colour =10 Hazen
GC/ECD (retention range 1,2,4-trichlorobenzene to decachlorobiphenyle, individual signals (lindane standard)) <= 3 pg/ml
GC/FID (retention range n-undecane to n-tetracontane, individual signals (n-tetradecane standard)) <= 3 ng/ml
Suitability Residue Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR