Aceton, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

1.00013.1000 1.00013.2500 1.00013.6010 1.00013.6025 1.00013.6190 1.00013.9950 1.00013.9180 1.00013.9911
1.00013.1000EA 93 EUR
1.00013.1000 1.00013.2500 1.00013.6010 1.00013.6025 1.00013.6190 1.00013.9950 1.00013.9180 1.00013.9911
Aceton, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Aceton
Formule: CH₃COCH₃
Kookpunt: 56,2 °C (1013 hPa)
Dichtheid: 0,792 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
Bereik: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable Veiligere gezondheid van mens en milieu Veiligere gezondheid van mens en milieu Veiligere gezondheid van mens en milieu
Milieu certificering Chem21 Solvent Selection Guide recommended after discussion Chem21 Solvent Selection Guide recommended after discussion Chem21 Solvent Selection Guide recommended after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR