Aceton, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-Propanone, DMK, Propanone, Dimethyl ketone

MQ300

1.00014.1000 1.00014.1011 1.00014.2500 1.00014.2511 1.00014.5000 1.00014.6010 1.00014.6025 1.00014.6190 1.00014.9180 1.00014.9900 1.00014.9912 1.00014.4000
1.00014.1000EA 22.77 EUR
1.00014.1000 1.00014.1011 1.00014.2500 1.00014.2511 1.00014.5000 1.00014.6010 1.00014.6025 1.00014.6190 1.00014.9180 1.00014.9900 1.00014.9912 1.00014.4000
Aceton, EMSURE® Reag. Ph. Eur., ACS, ISO analysis_for, Supelco®
Aceton

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. Merck declares their EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph. Eur.) and also with the ISO standards.

Formule: CH₃COCH₃
MW: 58.08 g/mol
Kookpunt: 56.2
Smeltpunt: –95.4 °C
Dichtheid: 0.792 g/cm³ (20 °C)
Vlampunt: <–20 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
Bereik: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -95.4 °C
Molar Mass 58.08 g/mol
Boiling Point 56.2 °C (1013 hPa)
Vapor Pressure 233 hPa (20 °C)
Refractive Index 1.35868 (20 °C)
Explosion Limit 2.6 - 12.8%(V)
Density 0.79 g/cm³ (20 °C)
pH 5 - 6 (395 g/L, H₂O, 20 °C)
Ignition Point 465 °C DIN 51794
Water Absorption 1000 g/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR