DL-Malic acid, Millipore®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid

M-Clarity™ quality level = MQ100

1.00382.0250 1.00382.1000
1.00382.0250EA 60.4 EUR
1.00382.0250 1.00382.1000
DL-Malic acid, Millipore®
DL-Malic acid
Formule: HOOCCH(OH)CH₂COOH
MW: 134.09 g/mol
Kookpunt: 306
Smeltpunt: 131…132 °C
Dichtheid: 1.601 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064212
CAS nummer: 617-48-1
EINECS: 230-022-8
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR