Oxaalzuur dihydraat, EMSURE® ISO, ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Ethanedioic acid dihydrate

M-Clarity™ quality level = MQ300

1.00495.0100 1.00495.0500 1.00495.1000 1.00495.9025
1.00495.0100EA 54.5 EUR
1.00495.0100 1.00495.0500 1.00495.1000 1.00495.9025
Oxaalzuur dihydraat, EMSURE® ISO, ACS, Reag. Ph. Eur. for analysis, Supelco®
Oxaalzuur dihydraat
Formule: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Kookpunt: 149…160 °C (1013 hPa)
Smeltpunt: 101
Dichtheid: 1.65 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149102
CAS nummer: 6153-56-6
EINECS: 205-634-3
UN: 3261
Bereik: 01-2119534576-33
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 102 g/l (20 °C)
Melting Point 101 °C
Molar Mass 126.07 g/mol
Boiling Point 149 - 160 °C (decomposition)
Vapor Pressure 21 hPa (50 °C)
Flash Point 157 °C (decomposition)
Density 1.65 g/cm3 (20 °C)
pH 1 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR