Testsets, Ozontest, voor Spectroquant®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.00607.0001 1.00607.0002
1.00607.0001EA 99.9 EUR
1.00607.0001 1.00607.0002
Testsets, Ozontest, voor Spectroquant®, Supelco®
Milieutestkits
Alle Spectroquant®-testkits kunnen worden gebruikt met de Prove spectrofotometers en Nova 60/60A-instrumenten. Tests kunnen niet alleen worden gebruikt met spectrofotometers van Merck, maar ook met fotometers en spectrofotometers van andere leveranciers (programmeerdetails op aanvraag beschikbaar).
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Order Now
Specificaties
Environmentally Preferable
M-Clarity quality level MQ100