Zwavelzuur 95-97%, EMSURE® ISO for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ300

1.00731.1000 1.00731.1011 1.00731.2500 1.00731.2511 1.00731.9025 1.00731.2510 1.00731.9201 1.00731.0510 1.00731.1010
1.00731.1000EA 37.5 EUR
1.00731.1000 1.00731.1011 1.00731.2500 1.00731.2511 1.00731.9025 1.00731.2510 1.00731.9201 1.00731.0510 1.00731.1010
Zwavelzuur 95-97%, EMSURE® ISO for analysis, Supelco®
Zwavelzuur
Formule: H₂SO₄
Kookpunt: ∼335 °C (1013 hPa)
Smeltpunt: 3 °C
Dichtheid: 1,84 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Solubility (20 °C) soluble,(caution ! development of heat)
Melting Point -20 °C
Molar Mass 98.08 g/mol
Boiling Point 335 °C
Vapor Pressure 0.0001 hPa (20 °C)
Density 1.84 g/cm3 (20 °C)
pH 0.3 (49 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR