Zirconium(IV) oxide, Technipur®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Zirconium dioxide, Zirconium Oxide

1.00757.9100
1.00757.9100EA 0 EUR
1.00757.9100
Zirconium(IV) oxide, Technipur®
Zirconium(IV) oxide
Formule: ZrO₂
Kookpunt: 4300 °C (1013 hPa)
Smeltpunt: 2680 °C
Dichtheid: 5,75 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011310
CAS nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR