L(+)-Tartaric acid, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, E334, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

1.00802.5000 1.00802.9029
1.00802.5000EA 1220 EUR
1.00802.5000 1.00802.9029
L(+)-Tartaric acid, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, E334, SAFC®
L(+)-Tartaric acid
Formule: C₄H₆O₆
Kookpunt: 399 °C (1013 hPa)
Smeltpunt: 168…170 °C
Dichtheid: 1,759 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064207
CAS nummer: 87-69-4
EINECS: 201-766-0
Bereik: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR