Ethanolamine, EMPROVE® ESSENTIAL BP, NF, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

1.00844.2500 1.00844.9025 1.00844.9180
1.00844.2500EA 198 EUR
1.00844.2500 1.00844.9025 1.00844.9180
Ethanolamine, EMPROVE® ESSENTIAL BP, NF, SAFC®
Ethanolamine
Formule: NH₂CH₂CH₂OH
Kookpunt: 171 °C (1013 hPa)
Dichtheid: 1,0164 g/cm³ (20 °C)
Vlampunt: 93 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008183
CAS nummer: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8,III
Merck Index: 13,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR