Ammoniumacetaat, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Acetic acid ammonium salt

M-Clarity™ quality level = MQ300

1.01116.0500_U 1.01116.1000 1.01116.5000 1.01116.9025 1.01116.9050 1.01116.9012
MERC1.01116.0500_UEA 111 EUR
MERC1.01116.0500_U 1.01116.1000 1.01116.5000 1.01116.9025 1.01116.9050 1.01116.9012
Ammoniumacetaat, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Ammoniumacetaat
Formule: H₃CCOONH₄
Smeltpunt: 114 °C
Dichtheid: 1,17 g/cm³ (20 °C)
Vlampunt: 136 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013066
CAS nummer: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1480 g/l (4 °C)
Melting Point 114 °C
Molar Mass 77.08 g/mol
Bulk Density 410 kg/m3
Flash Point 136 °C
Density 1.17 g/cm3 (20 °C)
pH 6.7 - 7.3 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR