Ammonium heptamolybdate tetrahydrate ACS, ISO analytical reagent, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate

M-Clarity™ quality level = MQ100

1.01182.0250 1.01182.1000 1.01182.5000
1.01182.0250EA 196.8 EUR
1.01182.0250 1.01182.1000 1.01182.5000
Ammonium heptamolybdate tetrahydrate ACS, ISO analytical reagent, Supelco®
Ammonium heptamolybdate tetrahydrate
Formule: (NH₄)₆Mo₇O₂₄·4H₂O
Dichtheid: 2,498 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00167059
CAS nummer: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Specification Test Results

Solubility 400 g/l (20 °C)
Melting Point 90 °C Elimination of water of crystallisation
Molar Mass 1235.86 g/mol
Bulk Density 800 kg/m3
Density 2.498 g/cm3 (20 °C)
pH 5.3 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR