Ammonium sulphate, Suprapur® for trace analysis, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

M-Clarity™ quality level = MQ100

1.01209.0050 1.01209.0500
1.01209.0050EA 92.82 EUR
1.01209.0050 1.01209.0500
Ammonium sulphate, Suprapur® for trace analysis, Supelco®
Ammonium sulphate
Formule: (NH₄)₂SO₄
Smeltpunt: 336…339 °C
Dichtheid: 1,78 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003391
CAS nummer: 7783-20-2
EINECS: 231-984-1
Bereik: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Solubility 754 g/l (20 °C)
Molar Mass 132.14 g/mol
Bulk Density 850 kg/m3
Density 1.77 g/cm3 (20 °C)
pH 5 (100 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR