Starch, soluble ISO analytical reagent, Millipore®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

1.01252.0100 1.01252.0250 1.01252.1000 1.01252.9025
1.01252.0100EA 62.2 EUR
1.01252.0100 1.01252.0250 1.01252.1000 1.01252.9025
Starch, soluble ISO analytical reagent, Millipore®
Starch, soluble
Formule: (C₆H₁₀O₅)n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00082026
CAS nummer: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR