Methyl orange Reag. Ph. Eur., ACS

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

M-Clarity™ quality level = MQ200

1.01322.0025 1.01322.0100 1.01322.1000
1.01322.0025EA 73.5 EUR
1.01322.0025 1.01322.0100 1.01322.1000
Methyl orange Reag. Ph. Eur., ACS
Methyl orange
Formule: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Kookpunt: ∼90 °C (1013 hPa)
Smeltpunt: > 300 °C
Dichtheid: ∼1 g/cm³ (20 °C)
Vlampunt: 38
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007502
CAS nummer: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1 III
Merck Index: 13,06127

Order Now

Specification Test Results

Solubility 5 g/l (20 °C)
Melting Point 300 °C
Molar Mass 327.34 g/mol
Bulk Density 200 - 400 kg/m3
pH 6.5 (5 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR