Barium sulphate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.01750.5000 1.01750.9050 1.01750.9025
1.01750.5000EA 593 EUR
1.01750.5000 1.01750.9050 1.01750.9025
Barium sulphate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Barium sulphate
Formule: BaSO₄
Kookpunt: 1600 °C (1013 hPa)
Smeltpunt: 1345 °C
Dichtheid: 4,5 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003455
CAS nummer: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR