Calcium thioglycolate trihydrate

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Calcium mercaptoacetate trihydrate, Calcium thioglycolate trihydrate, Calcium bis(mercaptoacetate) trihydrate, Mercaptoacetic acid calcium salt trihydrate

1.02125.1000 1.02125.9075
1.02125.1000EA 0 EUR
1.02125.1000 1.02125.9075
Calcium thioglycolate trihydrate
Calcium thioglycolate trihydrate
Formule: C₄H₆CaO₄S₂·3H₂O
MW: 276.35 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150707
CAS nummer: 65208-41-5
EINECS: 249-881-5
UN: 3263
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR