Cellulose for preparative chromatography
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.02331.0500 1.02331.2500 1.02331.9025
1.02331.0500EA 0 EUR
1.02331.0500 1.02331.2500 1.02331.9025
Cellulose for preparative chromatography
Sorbenten voor chromatografie Sorbenten voor HPLC
Micro-crystalline cellulose is a hydrophilic polysaccharide packing material preferred for the raw separation of amino acids and related compounds. Cellulose is also used for the gentle purification of bio-molecules. Because of the organic nature of cellulose, it can only be used in pre-swollen state and under gentle or hydrostatic pressure.
Order Now
Specificaties
Environmentally Preferable