Chloramine-T (sodium salt) trihydrate, extra pure, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

M-Clarity™ quality level = MQ200

1.02424.1000 1.02424.5000 1.02424.9050
1.02424.1000EA 392 EUR
1.02424.1000 1.02424.5000 1.02424.9050
Chloramine-T (sodium salt) trihydrate, extra pure, Supelco®
Chloramine-T (sodium salt) trihydrate
Formule: CH₃C₆H₄SO₂NClNa·3H₂O
Smeltpunt: 167…170 °C
Dichtheid: 1,43 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00149066
CAS nummer: 7080-50-4
EINECS: 204-854-7
UN: 2928
ADR: 6.1 (8),II
Merck Index: 13,02085

Order Now

Specification Test Results

Assay (iodometric) 98.0 - 103.0 %
Identity (IR-spectrum) passes test
Appearance of solution passes test
pH-value (5 %; water) 8.0 - 10.0
Matter insoluble in ethanol ≤ 2 %
ortho compounds passes test
Residual solvents (ICH Q3C) excluded by manufactoring process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR