Eosin Y (yellowish) 0,5% alcoholic solution Eosin Y-solution 0.5% alcoholic for microscopy, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

M-Clarity™ quality level = MQ300

1.02439.0500
1.02439.0500EA 56.4 EUR
1.02439.0500
Eosin Y (yellowish) 0,5% alcoholic solution Eosin Y-solution 0.5% alcoholic for microscopy, Sigma-Aldrich®
Eosin Y (yellowish)

Counterstain in H&E


Eosin Y solution 0.5% alcoholic, is a ready-to-use solution that can be used for routine staining in the hematoxylin-eosin staining process (H&E) of sample material of human origin. The H&E staining process is the most frequently used staining method for histological material. Due to the hematoxylin solution, the nuclei will appear blue to dark violet in color and the Eosin Y solution will stain the cytoplasma and intercellular substances red-orange.Sufficient for approx. 1500 specimens / liter. It is registered as IVD and CE product.

Formule: C₂₀H₆Br₄Na₂O₅
Dichtheid: 0,8889 g/cm3 (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005040
CAS nummer: 17372-87-1
UN: 1170
ADR: 3,II
IVDLogo

Order Now

Specification Test Results

Flash Point 19.1 °C
Density 0.8889 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR