Koper(II)acetaat monohydraat, crystals, EMPLURA®, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

M-Clarity™ quality level = MQ200

1.02710.0500 1.02710.9050 1.02710.1700
1.02710.0500EA 146 EUR
1.02710.0500 1.02710.9050 1.02710.1700
Koper(II)acetaat monohydraat, crystals, EMPLURA®, Supelco®
Koper(II)acetaat monohydraat
Formule: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Smeltpunt: 115
Dichtheid: 1.88 g/cm³ (20 °C)
MDL Nummer: MFCD00149570
CAS nummer: 6046-93-1
UN: 3077
ADR: 9 III
Merck Index: 13,02651

Order Now

Specification Test Results

Assay (iodometric) ≥ 99.0 %
In water insoluble matter ≤ 0.1 %
pH-value (2 %; water) 5.2 - 5.5
Chloride (Cl) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.2 %
Fe (Iron) ≤ 0.005 %
Zn (Zinc) ≤ 0.01 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR