Koper(II)oxide, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Cupric oxide, Copper oxide, Copper monoxide granulated

M-Clarity™ quality level = MQ200

1.02761.0500 1.02761.9025
1.02761.0500EA 714 EUR
1.02761.0500 1.02761.9025
Koper(II)oxide, Supelco®
Koper(II)oxide
Formule: CuO
MW: 79.55 g/mol
Kookpunt: 2000
Smeltpunt: 1326
Dichtheid: 4.9 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010979
CAS nummer: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 13,02674

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1326 °C (decomposition)
Molar Mass 79.55 g/mol
Bulk Density 500 kg/m3
Density 6.48 g/cm3 (25 °C)
pH 7 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR