L(+)-Cysteine, Millipore®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ100

1.02838.0025 1.02838.0100 1.02838.1000 1.02838.9010
1.02838.0025EA 51.7 EUR
1.02838.0025 1.02838.0100 1.02838.1000 1.02838.9010
L(+)-Cysteine, Millipore®
L-Cysteine
Formule: HSCH₂CH(NH₂)CO₂H
Kookpunt: 254 °C (1013 hPa)
Smeltpunt: 115 °C
Dichtheid: 1,523 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064306
CAS nummer: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR