Chloroform-[D₁] (99.8% D) stabilised, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

Stabilisatie: Stabilised with silver
M-Clarity™ quality level = MQ200

1.03420.0025 1.03420.0100 1.03420.0500
1.03420.0025EA 41.2 EUR
1.03420.0025 1.03420.0100 1.03420.0500
Chloroform-[D₁] (99.8% D) stabilised, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Chloroform-[D₁]
Formule: CDCl₃
Kookpunt: 60,8 °C (1013 hPa)
Dichtheid: 1,5 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000827
CAS nummer: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR